2,2,2-Trifluoro-1-(2,3,6-trifluorophenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

63264-500MG 63264-1G 63264-5G 63264-25G
77331-870EA 264.71 USD
77331-870 77356-168 77367-984 77389-330
2,2,2-Trifluoro-1-(2,3,6-trifluorophenyl)ethanone ≥97%
2,2,2-Trifluoro-1-(2,3,6-trifluorophenyl)ethanone

Precaución: for Research Use Only

Formula: C₈H₂F₆O
Storage Temperature: Ambient
MDL Number: MFCD12547804
CAS Number: 1208076-52-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR