Ethyl 3-bromo-5-iodo-2-methylbenzoate ≥95%

Supplier: AOBChem USA

Warning

63245-500MG 63245-1G 63245-5G 63245-25G
77331-868EA 202.91 USD
77331-868 77356-164 77367-982 77389-328
Ethyl 3-bromo-5-iodo-2-methylbenzoate ≥95%
Ethyl 3-bromo-5-iodo-2-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₀BrIO₂
Storage Temperature: Ambient
MDL Number: MFCD20526669
CAS Number: 1379317-96-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR