Methyl 3-bromo-5-iodo-2-methylbenzoate ≥95%

Supplier: AOBChem USA

Warning

63244-500MG 63244-1G 63244-5G 63244-25G
77331-866EA 147.29 USD
77331-866 77345-274 77367-980 77389-326
Methyl 3-bromo-5-iodo-2-methylbenzoate ≥95%
Methyl 3-bromo-5-iodo-2-methylbenzoate

Precaución: for Research Use Only

Formula: C₉H₈BrIO₂
Storage Temperature: Ambient
MDL Number: MFCD20526667
CAS Number: 1370600-52-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR