1-Isopropoxy-3-methyl-2-nitrobenzene ≥97%

Supplier: AOBChem USA

Warning

63242-500MG 63242-1G 63242-5G 63242-25G
77331-864EA 126.69 USD
77331-864 77344-186 77367-978 77380-198
1-Isopropoxy-3-methyl-2-nitrobenzene ≥97%
1-Isopropoxy-3-methyl-2-nitrobenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₃
MW: 195.2151 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29103861
CAS Number: 187471-35-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR