Ethyl 3-ethoxy-2-nitrobenzoate ≥97%

Supplier: AOBChem USA

Warning

63241-500MG 63241-1G 63241-5G 63241-25G
77331-862EA 202.91 USD
77331-862 77356-162 77367-976 77389-324
Ethyl 3-ethoxy-2-nitrobenzoate ≥97%
Ethyl 3-ethoxy-2-nitrobenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₃NO₅
MW: 239.2246 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01463644
CAS Number: 380635-88-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR