2-Amino-3-ethoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

63238-500MG 63238-1G 63238-25G 63238-5G
77331-858EA 292.52 USD
77331-858 77356-158 77389-320 77380-196
2-Amino-3-ethoxybenzoic acid ≥97%
2-Amino-3-ethoxybenzoic acid

Precaución: for Research Use Only

Formula: C₉H₁₁NO₃
MW: 181.1885 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12159989
CAS Number: 1015689-38-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR