Ethyl 2-chloro-6-ethoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

63223-1G 63223-5G 63223-25G
77340-792EA 60.77 USD
77340-792 77367-966 77380-192
Ethyl 2-chloro-6-ethoxybenzoate ≥97%
Ethyl 2-chloro-6-ethoxybenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₃ClO₃
MW: 228.6721 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761260
CAS Number: 2807445-55-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR