2-Chloro-1-methoxy-3-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

63214-500MG 63214-1G 63214-5G
77331-852EA 329.6 USD
77331-852 77356-152 77380-190
2-Chloro-1-methoxy-3-methylbenzene ≥97%
2-Chloro-1-methoxy-3-methylbenzene

Precaución: for Research Use Only

Formula: C₈H₉ClO
MW: 156.6094 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14705102
CAS Number: 90807-19-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR