4-Chloro-3-methoxy-5-methylbenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

63199-500MG 63199-1G 63199-5G
77338-722EA 511.91 USD
77338-722 77356-150 77380-188
4-Chloro-3-methoxy-5-methylbenzaldehyde ≥95%
4-Chloro-3-methoxy-5-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₉ClO₂
MW: 184.6195 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761251
CAS Number: 1785208-90-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR