2,4-Difluoro-5-methylaniline ≥97%

Supplier: AOBChem USA

Warning

63191-500MG 63191-1G 63191-5G 63191-25G
77331-848EA 272.95 USD
77331-848 77356-142 77367-962 77389-312
2,4-Difluoro-5-methylaniline ≥97%
2,4-Difluoro-5-methylaniline

Precaución: for Research Use Only

Formula: C₇H₇F₂N
Storage Temperature: Refrigerator
MDL Number: MFCD14583001
CAS Number: 1378579-56-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR