1-(4-Chloro-2,6-difluorophenyl)-2,2,2-trifluoroethanol ≥95%

Supplier: AOBChem USA

Warning

63177-500MG 63177-1G 63177-5G
77339-326EA 578.86 USD
77339-326 77356-136 77380-178
1-(4-Chloro-2,6-difluorophenyl)-2,2,2-trifluoroethanol ≥95%
1-(4-Chloro-2,6-difluorophenyl)-2,2,2-trifluoroethanol

Precaución: for Research Use Only

Formula: C₈H₄ClF₅O
Storage Temperature: Ambient
MDL Number: MFCD34761245
CAS Number: 2901098-58-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR