1-(4-Chloro-2,6-difluorophenyl)ethanol ≥97%

Supplier: AOBChem USA

Warning

63174-500MG 63174-1G 63174-5G 63174-25G
77331-844EA 223.51 USD
77331-844 77356-134 77367-960 77389-310
1-(4-Chloro-2,6-difluorophenyl)ethanol ≥97%
1-(4-Chloro-2,6-difluorophenyl)ethanol

Precaución: for Research Use Only

Formula: C₈H₇ClF₂O
Storage Temperature: Ambient
MDL Number: MFCD28523566
CAS Number: 1780642-82-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR