1-(3-Chloro-2,6-difluorophenyl)-2,2,2-trifluoroethanol ≥95%

Supplier: AOBChem USA

Warning

63147-500MG 63147-1G 63147-5G 63147-25G
77331-842EA 242.05 USD
77331-842 77356-132 77367-958 77389-308
1-(3-Chloro-2,6-difluorophenyl)-2,2,2-trifluoroethanol ≥95%
1-(3-Chloro-2,6-difluorophenyl)-2,2,2-trifluoroethanol

Precaución: for Research Use Only

Formula: C₈H₄ClF₅O
Storage Temperature: Ambient
MDL Number: MFCD32661975
CAS Number: 2229258-46-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR