1-(3-Chloro-2,6-difluorophenyl)-2,2,2-trifluoroethanone ≥97%

Supplier: AOBChem USA

Warning

63146-500MG 63146-1G 63146-5G
77336-226EA 383.16 USD
77336-226 77356-130 77380-176
1-(3-Chloro-2,6-difluorophenyl)-2,2,2-trifluoroethanone ≥97%
1-(3-Chloro-2,6-difluorophenyl)-2,2,2-trifluoroethanone

Precaución: for Research Use Only

Formula: C₈H₂ClF₅O
MW: 244.5458 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12922599
CAS Number: 1208077-28-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR