4-(Benzyloxy)-2-ethylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

63138-500MG 63138-1G 63138-5G
77331-838EA 348.14 USD
77331-838 77356-126 77380-172
4-(Benzyloxy)-2-ethylbenzoic acid ≥97%
4-(Benzyloxy)-2-ethylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₆H₁₆O₃
MW: 256.2964 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32184699
CAS Number: 851334-65-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR