4-(Benzyloxy)-2-ethylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

63136-500MG 63136-1G 63136-5G
77331-836EA 347.11 USD
77331-836 77356-124 77380-170
4-(Benzyloxy)-2-ethylbenzaldehyde ≥97%
4-(Benzyloxy)-2-ethylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₆H₁₆O₂
MW: 240.2970 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20484289
CAS Number: 668477-52-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR