1-(3-(Tert-butyl)phenyl)ethanol ≥95%

Supplier: AOBChem USA

Warning

63108-500MG 63108-1G 63108-5G 63108-25G
77331-830EA 243.08 USD
77331-830 77356-118 77367-956 77389-302
1-(3-(Tert-butyl)phenyl)ethanol ≥95%
1-(3-(Tert-butyl)phenyl)ethanol

Precaución: for Research Use Only

Formula: C₁₂H₁₈O
MW: 178.2707 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11520983
CAS Number: 71522-57-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR