(2-Chloro-5-iodophenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

63103-500MG 63103-1G 63103-5G 63103-25G
77331-828EA 134.93 USD
77331-828 77344-474 77367-954 77389-300
(2-Chloro-5-iodophenyl)(methyl)sulfane ≥97%
(2-Chloro-5-iodophenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₇H₆ClIS
MW: 284.5450 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761218
CAS Number: 2270843-54-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR