Ethyl 5-chloro-2-fluoro-4-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

63088-500MG 63088-1G 63088-5G 63088-25G
77331-824EA 154.5 USD
77331-824 77345-554 77367-950 77389-296
Ethyl 5-chloro-2-fluoro-4-methylbenzoate ≥97%
Ethyl 5-chloro-2-fluoro-4-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₀ClFO₂
MW: 216.6366 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24108607
CAS Number: 1538528-61-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR