Ethyl 3-(benzyloxy)-2-fluoro-4-methoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

63083-500MG 63083-1G 63083-5G
77337-628EA 431.57 USD
77337-628 77356-112 77380-164
Ethyl 3-(benzyloxy)-2-fluoro-4-methoxybenzoate ≥97%
Ethyl 3-(benzyloxy)-2-fluoro-4-methoxybenzoate

Precaución: for Research Use Only

Formula: C₁₇H₁₇FO₄
MW: 304.3128 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761212
CAS Number: 2807440-26-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR