(3-(Benzyloxy)-2-fluoro-4-methoxyphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

63080-500MG 63080-1G 63080-5G
77337-790EA 443.93 USD
77337-790 77356-108 77380-160
(3-(Benzyloxy)-2-fluoro-4-methoxyphenyl)methanol ≥97%
(3-(Benzyloxy)-2-fluoro-4-methoxyphenyl)methanol

Precaución: for Research Use Only

Formula: C₁₅H₁₅FO₃
MW: 262.2762 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761209
CAS Number: 2807466-31-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR