2-(Benzyloxy)-3-fluoro-4-iodo-1-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

63077-500MG 63077-1G 63077-5G 63077-25G
77331-820EA 258.53 USD
77331-820 77356-104 77367-946 77389-292
2-(Benzyloxy)-3-fluoro-4-iodo-1-methoxybenzene ≥97%
2-(Benzyloxy)-3-fluoro-4-iodo-1-methoxybenzene

Precaución: for Research Use Only

Formula: C₁₄H₁₂FIO₂
MW: 358.1467 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761206
CAS Number: 2807443-07-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR