3-Ethoxy-2-fluoro-4-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

63070-500MG 63070-1G 63070-5G 63070-25G
77331-816EA 126.69 USD
77331-816 77344-220 77367-942 77380-156
3-Ethoxy-2-fluoro-4-methoxybenzoic acid ≥97%
3-Ethoxy-2-fluoro-4-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁FO₄
Storage Temperature: Ambient
MDL Number: MFCD34761199
CAS Number: 2387058-65-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR