1-Fluoro-2-isopropoxy-3-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

63067-1G 63067-5G 63067-25G
77341-248EA 78.28 USD
77341-248 77367-940 77380-154
1-Fluoro-2-isopropoxy-3-methoxybenzene ≥97%
1-Fluoro-2-isopropoxy-3-methoxybenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₃FO₂
Storage Temperature: Ambient
MDL Number: MFCD23114329
CAS Number: 1369779-10-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR