1-Bromo-2-fluoro-3-isopropoxy-4-methoxybenzene ≥95%

Supplier: AOBChem USA

Warning

63065-500MG 63065-1G 63065-5G 63065-25G
77331-814EA 188.49 USD
77331-814 77356-100 77367-938 77389-288
1-Bromo-2-fluoro-3-isopropoxy-4-methoxybenzene ≥95%
1-Bromo-2-fluoro-3-isopropoxy-4-methoxybenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₂BrFO₂
MW: 263.1034 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29043901
CAS Number: 1881293-79-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR