2-(Benzyloxy)-4-bromo-3-fluoro-1-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

63064-500MG 63064-1G 63064-5G 63064-25G
77331-812EA 184.37 USD
77331-812 77347-256 77367-936 77389-286
2-(Benzyloxy)-4-bromo-3-fluoro-1-methoxybenzene ≥97%
2-(Benzyloxy)-4-bromo-3-fluoro-1-methoxybenzene

Precaución: for Research Use Only

Formula: C₁₄H₁₂BrFO₂
MW: 311.1462 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761197
CAS Number: 1713253-20-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR