2,3,4-Trifluoro-5-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

63059-500MG 63059-1G 63059-5G 63059-25G
77331-810EA 128.75 USD
77331-810 77344-280 77367-934 77380-150
2,3,4-Trifluoro-5-methylbenzaldehyde ≥97%
2,3,4-Trifluoro-5-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₅F₃O
Storage Temperature: Ambient
MDL Number: MFCD34761194
CAS Number: 866925-67-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR