4-(Benzyloxy)-3-bromo-5-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

63034-1G 63034-5G 63034-25G
77340-978EA 65.92 USD
77340-978 77367-932 77380-148
4-(Benzyloxy)-3-bromo-5-methoxybenzaldehyde ≥97%
4-(Benzyloxy)-3-bromo-5-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₅H₁₃BrO₃
Storage Temperature: Ambient
MDL Number: MFCD01119124
CAS Number: 2556-04-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR