3-Fluoro-5-(hydroxymethyl)phenol ≥97%

Supplier: AOBChem USA

Warning

63033-500MG 63033-1G 63033-5G
77336-224EA 383.16 USD
77336-224 77356-098 77380-146
3-Fluoro-5-(hydroxymethyl)phenol ≥97%
3-Fluoro-5-(hydroxymethyl)phenol

Precaución: for Research Use Only

Formula: C₇H₇FO₂
MW: 142.1276 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16249597
CAS Number: 1260762-16-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR