4-Bromo-2-fluoro-3,4'-dimethyl-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

63032-500MG 63032-1G 63032-5G
77339-320EA 577.83 USD
77339-320 77356-096 77380-144
4-Bromo-2-fluoro-3,4'-dimethyl-1,1'-biphenyl ≥97%
4-Bromo-2-fluoro-3,4'-dimethyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₂BrF
MW: 279.1474 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761190
CAS Number: 2764732-17-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR