4-Bromo-2,4'-difluoro-3-methyl-1,1'-biphenyl ≥95%

Supplier: AOBChem USA

Warning

63030-500MG 63030-1G 63030-5G
77338-788EA 517.06 USD
77338-788 77356-094 77380-142
4-Bromo-2,4'-difluoro-3-methyl-1,1'-biphenyl ≥95%
4-Bromo-2,4'-difluoro-3-methyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₃H₉BrF₂
MW: 283.1113 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761189
CAS Number: 2764728-19-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR