Methyl 3-(benzyloxy)-5-chlorobenzoate ≥97%

Supplier: AOBChem USA

Warning

63018-500MG 63018-1G 63018-5G 63018-25G
77331-808EA 214.24 USD
77331-808 77356-092 77367-930 77389-284
Methyl 3-(benzyloxy)-5-chlorobenzoate ≥97%
Methyl 3-(benzyloxy)-5-chlorobenzoate

Precaución: for Research Use Only

Formula: C₁₅H₁₃ClO₃
MW: 276.7149 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761178
CAS Number: 1098592-80-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR