Methyl 2-(methylthio)thiophene-3-carboxylate ≥97%

Supplier: AOBChem USA

Warning

63016-500MG 63016-1G 63016-5G
77331-804EA 319.3 USD
77331-804 77356-088 77380-140
Methyl 2-(methylthio)thiophene-3-carboxylate ≥97%
Methyl 2-(methylthio)thiophene-3-carboxylate

Precaución: for Research Use Only

Formula: C₇H₈O₂S₂
MW: 188.2672 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06204148
CAS Number: 76360-40-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR