(2-Fluoro-3,5-dimethylphenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

63005-500MG 63005-1G 63005-5G 63005-25G
77331-802EA 161.71 USD
77331-802 77345-916 77367-926 77389-280
(2-Fluoro-3,5-dimethylphenyl)methanol ≥95%
(2-Fluoro-3,5-dimethylphenyl)methanol

Precaución: for Research Use Only

Formula: C₉H₁₁FO
MW: 154.1814 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28551042
CAS Number: 1785361-54-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR