(3-Bromo-5-chloro-2-methylphenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

62996-500MG 62996-1G 62996-5G
77339-508EA 594.31 USD
77339-508 77356-086 77380-138
(3-Bromo-5-chloro-2-methylphenyl)boronic acid ≥97%
(3-Bromo-5-chloro-2-methylphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₇H₇BBrClO₂
Storage Temperature: Ambient
MDL Number: MFCD24449157
CAS Number: 2225155-54-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR