3-Bromo-5-chloro-2-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

62984-500MG 62984-1G 62984-5G
77340-326EA 900.22 USD
77340-326 77356-084 77380-136
3-Bromo-5-chloro-2-methylbenzaldehyde ≥97%
3-Bromo-5-chloro-2-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆BrClO
MW: 233.4896 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28396363
CAS Number: 1934421-65-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR