1-Bromo-5-chloro-3-iodo-2-methylbenzene ≥97%
Supplier: AOBChem USA
|
Warning
|
62977-500MG
62977-1G
62977-5G
77336-116EA
375.95
USD
77336-116
77356-082
77380-134
1-Bromo-5-chloro-3-iodo-2-methylbenzene ≥97%
1-Bromo-5-chloro-3-iodo-2-methylbenzene
Precaución: for Research Use Only
Formula:
C₇H₅BrClI MW: 331.3760 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34761164 CAS Number: 2385789-84-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...