(3-Bromo-5-chloro-2-methylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

62976-500MG 62976-1G 62976-5G
77331-800EA 347.11 USD
77331-800 77356-080 77380-132
(3-Bromo-5-chloro-2-methylphenyl)(methyl)sulfane ≥97%
(3-Bromo-5-chloro-2-methylphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₈H₈BrClS
MW: 251.5711 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761163
CAS Number: 2710184-20-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR