(5-Bromo-[1,1'-biphenyl]-3-yl)methanol ≥97%

Supplier: AOBChem USA

Warning

62946-500MG 62946-1G 62946-5G 62946-25G
77331-796EA 229.69 USD
77331-796 77356-074 77367-924 77389-278
(5-Bromo-[1,1'-biphenyl]-3-yl)methanol ≥97%
(5-Bromo-[1,1'-biphenyl]-3-yl)methanol

Precaución: for Research Use Only

Formula: C₁₃H₁₁BrO
MW: 263.1298 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26959968
CAS Number: 1823881-88-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR