1-Chloro-5-ethoxy-2-iodo-4-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

62897-500MG 62897-1G 62897-5G
77337-316EA 410.97 USD
77337-316 77356-070 77380-124
1-Chloro-5-ethoxy-2-iodo-4-methylbenzene ≥97%
1-Chloro-5-ethoxy-2-iodo-4-methylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₀ClIO
MW: 296.5325 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761110
CAS Number: 2385261-67-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR