(2-Chloro-4-ethoxy-5-methylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

62896-500MG 62896-1G 62896-5G
77339-768EA 633.45 USD
77339-768 77356-068 77380-122
(2-Chloro-4-ethoxy-5-methylphenyl)(methyl)sulfane ≥97%
(2-Chloro-4-ethoxy-5-methylphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₁₀H₁₃ClOS
Storage Temperature: Ambient
MDL Number: MFCD34761109
CAS Number: 2705764-69-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR