1-(2-Chloro-4-ethoxy-5-methylphenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

62892-500MG 62892-1G 62892-5G
77338-428EA 487.19 USD
77338-428 77356-066 77380-120
1-(2-Chloro-4-ethoxy-5-methylphenyl)ethanone ≥97%
1-(2-Chloro-4-ethoxy-5-methylphenyl)ethanone

Precaución: for Research Use Only

Formula: C₁₁H₁₃ClO₂
MW: 212.6727 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761105
CAS Number: 2386338-84-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR