(2-Chloro-4-methoxy-5-methylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

62890-500MG 62890-1G 62890-5G
77338-786EA 517.06 USD
77338-786 77356-064 77380-118
(2-Chloro-4-methoxy-5-methylphenyl)(methyl)sulfane ≥97%
(2-Chloro-4-methoxy-5-methylphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₉H₁₁ClOS
MW: 202.7010 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761104
CAS Number: 2708689-79-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR