1-Cyclobutyl-2,3-difluorobenzene ≥97%

Supplier: AOBChem USA

Warning

62866-500MG 62866-1G 62866-5G
77338-536EA 496.46 USD
77338-536 77356-062 77380-116
1-Cyclobutyl-2,3-difluorobenzene ≥97%
1-Cyclobutyl-2,3-difluorobenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₀F₂
MW: 168.1832 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761086
CAS Number: 2901082-81-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR