1-(2,3-Difluorophenyl)cyclobutanol ≥97%

Supplier: AOBChem USA

Warning

62865-500MG 62865-1G 62865-5G 62865-25G
77331-794EA 202.91 USD
77331-794 77356-060 77367-922 77389-276
1-(2,3-Difluorophenyl)cyclobutanol ≥97%
1-(2,3-Difluorophenyl)cyclobutanol

Precaución: for Research Use Only

Formula: C₁₀H₁₀F₂O
MW: 184.1826 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21788678
CAS Number: 1514553-81-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR