3-Fluoro-4-formyl-5-methoxybenzoic acid ≥95%

Supplier: AOBChem USA

Warning

62855-500MG 62855-1G
77340-512EA 1371.96 USD
77340-512 77356-056
3-Fluoro-4-formyl-5-methoxybenzoic acid ≥95%
3-Fluoro-4-formyl-5-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₉H₇FO₄
Storage Temperature: Ambient
MDL Number: MFCD34761077
CAS Number: 2387218-10-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR