2-Fluoro-6-methoxy-4-(methylthio)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

62853-500MG 62853-1G 62853-5G
77339-504EA 594.31 USD
77339-504 77356-054 77380-112
2-Fluoro-6-methoxy-4-(methylthio)benzaldehyde ≥97%
2-Fluoro-6-methoxy-4-(methylthio)benzaldehyde

Precaución: for Research Use Only

Formula: C₉H₉FO₂S
MW: 200.2299 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761075
CAS Number: 2807469-57-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR