2-Fluoro-4-iodo-6-methoxybenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

62851-500MG 62851-1G 62851-5G
77340-372EA 965.11 USD
77340-372 77356-052 77380-110
2-Fluoro-4-iodo-6-methoxybenzaldehyde ≥95%
2-Fluoro-4-iodo-6-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆FIO₂
Storage Temperature: Ambient
MDL Number: MFCD32875368
CAS Number: 2090466-20-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR