2,3-Dichloro-5-fluorophenol ≥97%

Supplier: AOBChem USA

Warning

62843-500MG 62843-1G 62843-5G
77338-542EA 497.49 USD
77338-542 77356-048 77380-108
2,3-Dichloro-5-fluorophenol ≥97%
2,3-Dichloro-5-fluorophenol

Precaución: for Research Use Only

Formula: C₆H₃Cl₂FO
MW: 180.9918 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28786272
CAS Number: 1804515-85-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR