2,3-Dichloro-5-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

62837-500MG 62837-1G 62837-5G
77331-788EA 321.36 USD
77331-788 77356-046 77380-106
2,3-Dichloro-5-methoxybenzoic acid ≥97%
2,3-Dichloro-5-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆Cl₂O₃
Storage Temperature: Ambient
MDL Number: MFCD34761066
CAS Number: 2091216-81-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR